Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npnt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001287101.1 | NP_001274030.1 | 5 - 6 | 653 | 61 | |
NM_001287102.1 | NP_001274031.1 | 4 - 5 | 602 | 61 | ||
GenBank mRNA | AF397007.1 | - | 4 - 5 | 354 | 61 | |
AF397008.1 | - | 5 - 6 | 405 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |