Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc4a10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001242378.1 | NP_001229307.1 | 3 - 4 | 404 | 85 | |
NM_001242379.1 | NP_001229308.1 | 3 - 4 | 404 | 85 | ||
NM_001242380.1 | NP_001229309.1 | 3 - 4 | 404 | 85 | ||
NM_001242381.1 | NP_001229310.1 | 3 - 4 | 404 | 85 | ||
NM_001242382.1 | NP_001229311.1 | 3 - 4 | 404 | 85 | ||
NM_001242383.1 | NP_001229312.1 | 3 - 4 | 404 | 85 | ||
NM_033552.3 | NP_291030.2 | 3 - 4 | 404 | 85 | ||
GenBank mRNA | AB033759.1 | - | 3 - 4 | 325 | 85 | |
AK043017.1 | - | 3 - 4 | 380 | 85 | ||
AK220501.1 | - | 3 - 4 | 369 | 85 | ||
BC039226.1 | - | 3 - 4 | 356 | 4480841 | 85 | |
CF746204.1 | - | 3 - 4 | 404 | 85 | ||
HM149278.1 | - | 3 - 4 | 365 | 85 | ||
HQ533147.1 | - | 3 - 4 | 344 | 85 | ||
HQ825316.1 | - | 3 - 4 | 344 | 85 | ||
HQ831517.1 | - | 3 - 4 | 344 | 85 | ||
JF500487.1 | - | 3 - 4 | 481 | 85 | ||
JF500488.1 | - | 3 - 4 | 481 | 85 | ||
JF500489.1 | - | 3 - 4 | 481 | 85 | ||
JF500490.1 | - | 3 - 4 | 481 | 85 | ||
JX220977.1 | - | 3 - 4 | 371 | 85 | ||
JX254917.1 | - | 3 - 4 | 371 | 85 | ||
JX268544.1 | - | 3 - 4 | 482 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |