Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arid1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080819.1 | NP_001074288.1 | 18 - 19 | 4993 | 72 | |
GenBank mRNA | AF268912.1 | - | 17 - 18 | 3856 | 72 | |
BC082554.1 | - | 4237 | 30622048 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |