Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dach2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142570.1 | NP_001136042.1 | 4 - 5 | 1087 | 98 | |
NM_001289732.1 | NP_001276661.1 | 7 - 8 | 886 | 98 | ||
NM_001289733.1 | NP_001276662.1 | 6 - 7 | 804 | 98 | ||
NM_001289734.1 | NP_001276663.1 | 7 - 8 | 955 | 98 | ||
NM_033605.2 | NP_291083.1 | 5 - 6 | 1126 | 98 | ||
GenBank mRNA | AF257217.1 | - | 5 - 6 | 1125 | 98 | |
AK033050.1 | - | 7 - 8 | 887 | 98 | ||
AK041446.1 | - | 6 - 7 | 793 | 98 | ||
AK042401.1 | - | 7 - 8 | 890 | 98 | ||
AK045154.1 | - | 5 - 6 | 1129 | 98 | ||
BC059233.1 | - | 4 - 5 | 1076 | 6406445 | 98 | |
GQ247884.1 | - | 2 - 3 | 265 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |