Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ophn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_052976.3 | NP_443208.1 | 19 - 20 | 2305 | 100 | |
GenBank mRNA | AK028822.1 | - | 19 - 20 | 1789 | 100 | |
AK031419.1 | - | 19 - 20 | 2298 | 100 | ||
AK036038.1 | - | 19 - 20 | 2306 | 100 | ||
AK082551.1 | - | 19 - 20 | 1945 | 100 | ||
AK087469.1 | - | 19 - 20 | 1769 | 100 | ||
BC004845.1 | - | 20 - 21 | 1903 | 2650009 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |