Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ophn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_052976.3 | NP_443208.1 | 21 - 22 | 2784 | 62 | |
GenBank mRNA | AK028822.1 | - | 21 - 22 | 2268 | 62 | |
AK031419.1 | - | 21 - 22 | 2777 | 62 | ||
AK036038.1 | - | 21 - 22 | 2785 | 62 | ||
AK082551.1 | - | 21 - 22 | 2424 | 62 | ||
AK087469.1 | - | 21 - 22 | 2248 | 62 | ||
BC004845.1 | - | 22 - 23 | 2382 | 2650009 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |