Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adarb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289530.1 | NP_001276459.1 | 7 - 8 | 2053 | 69 | |
NM_052977.5 | NP_443209.2 | 7 - 8 | 2053 | 69 | ||
NR_110345.1 | 7 - 8 | 2053 | 69 | |||
GenBank mRNA | AF232942.1 | - | 7 - 8 | 1670 | 69 | |
AF270495.1 | - | 7 - 8 | 1770 | 69 | ||
AK033916.1 | - | 7 - 8 | 1992 | 69 | ||
AK045538.1 | - | 7 - 8 | 1993 | 69 | ||
AK046083.1 | - | 7 - 8 | 1997 | 69 | ||
AK134964.1 | - | 7 - 8 | 2055 | 69 | ||
AK139079.1 | - | 7 - 8 | 2011 | 69 | ||
AK139273.1 | - | 7 - 8 | 2011 | 69 | ||
BC052426.1 | - | 7 - 8 | 1978 | 6835942 | 69 | |
BC059822.1 | - | 7 - 8 | 2006 | 6844585 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |