Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400007943.mm.1 |
NM_001080926.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400007943.mm.1 |
NM_001080926.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400007943.mm.1 |
NR_033496.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400007943.mm.1 |
NR_033496.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400007943.mm.1 |
TC0400001135.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400007941.mm.1 |
NM_001080926.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400007941.mm.1 |
NM_001080926.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400007941.mm.1 |
NR_033496.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400007941.mm.1 |
NR_033496.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400007941.mm.1 |
TC0400001135.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400007942.mm.1 |
NM_001080926.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400007942.mm.1 |
NM_001080926.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400007942.mm.1 |
NR_033496.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400007942.mm.1 |
NR_033496.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400007942.mm.1 |
TC0400001135.mm.1 |
MTA TC |