Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldh1a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053080.3 | NP_444310.3 | 9 - 10 | 1137 | 71 | |
GenBank mRNA | AF152359.1 | - | 9 - 10 | 1092 | 71 | |
AF246711.1 | - | 9 - 10 | 1085 | 71 | ||
AF253409.1 | - | 9 - 10 | 1124 | 71 | ||
AF280404.1 | - | 9 - 10 | 1089 | 71 | ||
AK147006.1 | - | 9 - 10 | 1138 | 71 | ||
BC048941.1 | - | 1 - 2 | 50 | 71 | ||
BC058277.1 | - | 9 - 10 | 1081 | 6515355 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |