Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tac4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053093.1 | NP_444323.1 | 2 - 3 | 390 | 75 | |
GenBank mRNA | AF235035.1 | - | 2 - 3 | 390 | 75 | |
AF515827.1 | - | 2 - 3 | 224 | 75 | ||
BC119426.1 | - | 2 - 3 | 370 | 40039772 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |