Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ces1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053200.2 | NP_444430.2 | 6 - 7 | 886 | 101 | |
GenBank mRNA | AB023631.1 | - | 6 - 7 | 846 | 101 | |
AF378751.1 | - | 6 - 7 | 861 | 101 | ||
AK078879.1 | - | 6 - 7 | 864 | 101 | ||
BC019198.1 | - | 6 - 7 | 858 | 5040523 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |