Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ehbp1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114597.1 | NP_001108069.1 | 9 - 10 | 3909 | 153 | |
GenBank mRNA | AF305087.1 | - | 9 - 10 | 3865 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons |