Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc13a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_054055.2 | NP_473396.1 | 2 - 3 | 413 | 89 | |
GenBank mRNA | AF306491.1 | - | 2 - 3 | 412 | 89 | |
AK085326.1 | - | 2 - 3 | 411 | 89 | ||
AK086775.1 | - | 2 - 3 | 415 | 89 | ||
AK087518.1 | - | 2 - 3 | 411 | 89 | ||
AK128982.1 | - | 2 - 3 | 410 | 89 | ||
AK138344.1 | - | 2 - 3 | 411 | 89 | ||
AK157881.1 | - | 2 - 3 | 382 | 89 | ||
AK165355.1 | - | 2 - 3 | 411 | 89 | ||
BC026803.1 | - | 2 - 3 | 399 | 4238769 | 89 | |
BC028882.1 | - | 2 - 3 | 397 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |