Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2180904 |
AK153750.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2180904 |
BC014874.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2180904 |
AF333980.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2180904 |
NM_054076.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2180904 |
NM_001160420.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
473229 |
BC099490.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
473229 |
NM_001025606.1 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
739738 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
868007 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
297281 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
535539 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
497849 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
9416 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P161539 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2180904 |
AK153750.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2180904 |
BC014874.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2180904 |
AF333980.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2180904 |
NM_054076.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2180904 |
NM_001160420.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20630219 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20630226 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20630234 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20630205 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20630227 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20630214 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20630206 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20630209 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20630211 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20630238 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20630225 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20630229 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20630240 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20630222 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20630217 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20630215 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20630232 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20630237 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20630208 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20630231 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P161539 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P04270530 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P241306 |
AF333980.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P113026 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P161539 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P00518744 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P00863657 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P00863656 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P00863662 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P00518747 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20630117 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20630289 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20630215 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20630291 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20630088 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20630217 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20630169 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20630119 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20630193 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20630203 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20630294 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20630091 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20630114 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20630179 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20630234 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20630286 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20630293 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20630102 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20630156 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20630132 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20630287 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20630191 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20630092 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20630184 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20630205 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20630197 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20630231 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20630142 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20630284 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20630190 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20630137 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20630225 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20630200 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20630209 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20630230 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20630087 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20630177 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20630157 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20630208 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20630101 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20630226 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20630154 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20630195 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20630126 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20630164 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20630280 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20630116 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20630196 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20630097 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20630148 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20630089 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20630162 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20630181 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20630159 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20630095 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20630149 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20630112 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20630174 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20630283 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20630206 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20630113 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20630292 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20630278 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20630152 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20630238 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20630201 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20630129 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20630130 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20630123 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20630111 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20630172 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20630106 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20630135 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20630138 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20630211 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20630170 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20630108 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20630146 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20630186 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20630124 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20630290 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20630222 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20630240 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01089767 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00546660 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20630138 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00518752 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20630206 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00196715 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00518751 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20630106 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20630134 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20630125 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20630164 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00196719 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20630205 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00546658 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00398176 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01089763 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00975793 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01056227 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20630102 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00149378 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00696773 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01089729 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20630196 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20630201 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20630230 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01089748 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01089759 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01089746 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20630124 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20630117 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01089737 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20630203 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20630220 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00518747 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20630126 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00866177 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20630097 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00616168 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00863667 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00149379 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00149382 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01089747 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20630094 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20630099 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20630225 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20630101 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20630108 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00975794 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20630113 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20630226 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20630141 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01089726 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00546655 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00398173 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01089768 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20630219 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00149364 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20630129 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00196720 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01089760 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00546656 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20630166 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00196711 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00863658 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00975796 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20630123 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20630162 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20630222 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00744076 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20630092 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01056229 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20630132 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01089727 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00149374 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20630177 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20630169 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20630240 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00149377 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20630241 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20630167 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20630170 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20630146 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00149370 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20630140 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00289554 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20630130 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00196714 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01056230 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00518753 |
- |
- |