Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prelp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_054077.4 | NP_473418.3 | 1 - 2 | 143 | 81 | |
GenBank mRNA | AK037150.1 | - | 1 - 2 | 121 | 81 | |
AK047480.1 | - | 1 - 2 | 144 | 81 | ||
BC019775.1 | - | 1 - 2 | 119 | 4165772 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |