Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Necab2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_054095.2 | NP_473436.1 | 9 - 10 | 1191 | 68 | |
GenBank mRNA | AF411253.1 | - | 9 - 10 | 1185 | 68 | |
AK035168.1 | - | 7 - 8 | 564 | 68 | ||
AK082727.1 | - | 9 - 10 | 801 | 68 | ||
BC107384.1 | - | 10 - 11 | 786 | 68 | ||
BC107385.1 | - | 10 - 11 | 750 | 68 | ||
BC115857.1 | - | 10 - 11 | 723 | 40057629 | 68 | |
BC115963.1 | - | 10 - 11 | 711 | 40057628 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |