Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lpo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC016212.1 | - | 4 - 5 | 708 | 4924899 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |