Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007400.2 | NP_031426.2 | 12 - 13 | 1544 | 63 | |
GenBank mRNA | AK051780.1 | - | 2 - 3 | 358 | 63 | |
D50411.1 | - | 12 - 13 | 1544 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mus musculus Probe spans exons |