Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007400.2 | NP_031426.2 | 19 - 20 | 2459 | 96 | |
GenBank mRNA | AK051780.1 | - | 9 - 10 | 1273 | 96 | |
AK220274.1 | - | 6 - 7 | 669 | 96 | ||
D50411.1 | - | 19 - 20 | 2459 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease Mus musculus Probe spans exons |