Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aup1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001301649.1 | NP_001288578.1 | 1 - 2 | 315 | 75 | |
NM_007517.4 | NP_031543.2 | 1 - 2 | 315 | 75 | ||
NR_125891.1 | 1 - 2 | 315 | 75 | |||
GenBank mRNA | AK154707.1 | - | 1 - 2 | 184 | 75 | |
AK158035.1 | - | 1 - 2 | 106 | 75 | ||
AK167335.1 | - | 1 - 2 | 105 | 75 | ||
AK171011.1 | - | 1 - 2 | 118 | 75 | ||
AK171184.1 | - | 1 - 2 | 110 | 75 | ||
AK171202.1 | - | 1 - 2 | 124 | 75 | ||
AK171249.1 | - | 1 - 2 | 104 | 75 | ||
AK171541.1 | - | 1 - 2 | 105 | 75 | ||
AK171621.1 | - | 1 - 2 | 96 | 75 | ||
BC012699.1 | - | 1 - 2 | 99 | 3986517 | 75 | |
BC016485.1 | - | 1 - 2 | 108 | 3962982 | 75 | |
U41736.1 | - | 1 - 2 | 113 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Probe spans exons |