Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Blm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042527.2 | NP_001035992.1 | 5 - 6 | 1183 | 71 | |
NM_007550.4 | NP_031576.4 | 5 - 6 | 1454 | 71 | ||
GenBank mRNA | AB008674.1 | - | 5 - 6 | 1447 | 71 | |
AK137022.1 | - | 5 - 6 | 1165 | 71 | ||
AK157898.1 | - | 5 - 6 | 1187 | 71 | ||
AK164605.1 | - | 5 - 6 | 1152 | 71 | ||
Z98263.1 | - | 5 - 6 | 1161 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Inventoried Probe spans exons |