Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nedd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008682.2 | NP_032708.2 | 5 - 6 | 593 | 92 | |
GenBank mRNA | AK010591.1 | - | 5 - 6 | 581 | 92 | |
AK048393.1 | - | 5 - 6 | 545 | 92 | ||
AK085355.1 | - | 5 - 6 | 556 | 92 | ||
AK090040.1 | - | 5 - 6 | 538 | 92 | ||
AK153746.1 | - | 5 - 6 | 595 | 92 | ||
BC052430.1 | - | 5 - 6 | 562 | 6836583 | 92 | |
BC066870.1 | - | 5 - 6 | 530 | 30476383 | 92 | |
D10712.1 | - | 5 - 6 | 535 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |