Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pafah1b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008775.3 | NP_032801.2 | 2 - 3 | 255 | 90 | |
GenBank mRNA | AK012116.1 | - | 2 - 3 | 169 | 90 | |
AK012164.1 | - | 2 - 3 | 190 | 90 | ||
AK153424.1 | - | 2 - 3 | 226 | 90 | ||
BB856076.1 | - | 2 - 3 | 256 | 90 | ||
BC002037.1 | - | 2 - 3 | 203 | 3486003 | 90 | |
BC056211.2 | - | 2 - 3 | 225 | 30012448 | 90 | |
U57747.1 | - | 2 - 3 | 187 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |