Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcolce | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008788.2 | NP_032814.2 | 6 - 7 | 1085 | 106 | |
GenBank mRNA | AB008548.1 | - | 6 - 7 | 1009 | 106 | |
AK142615.1 | - | 6 - 7 | 1014 | 106 | ||
AK144392.1 | - | 6 - 7 | 1086 | 106 | ||
AK146829.1 | - | 6 - 7 | 1086 | 106 | ||
AK159430.1 | - | 6 - 7 | 1073 | 106 | ||
AK159940.1 | - | 6 - 7 | 1077 | 106 | ||
X57337.1 | - | 6 - 7 | 1052 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |