Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcsk7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001281934.1 | NP_001268863.1 | 7 - 8 | 1781 | 64 | |
NM_008794.2 | NP_032820.2 | 7 - 8 | 1517 | 64 | ||
NR_104055.1 | 7 - 8 | 1517 | 64 | |||
GenBank mRNA | AK148416.1 | - | 7 - 8 | 1102 | 64 | |
AK151980.1 | - | 7 - 8 | 1104 | 64 | ||
AK165389.1 | - | 7 - 8 | 1123 | 64 | ||
BC006730.1 | - | 6 - 7 | 1018 | 3962441 | 64 | |
BC007322.1 | - | 7 - 8 | 1380 | 64 | ||
BC056456.1 | - | 7 - 8 | 1517 | 64 | ||
U48830.1 | - | 7 - 8 | 1089 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Mus musculus Probe spans exons |