Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcsk7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001281934.1 | NP_001268863.1 | 10 - 11 | 2194 | 87 | |
NM_008794.2 | NP_032820.2 | 10 - 11 | 1930 | 87 | ||
NR_104055.1 | 10 - 11 | 1930 | 87 | |||
GenBank mRNA | AK148416.1 | - | 10 - 11 | 1515 | 87 | |
AK151980.1 | - | 10 - 11 | 1517 | 87 | ||
AK165389.1 | - | 10 - 11 | 1536 | 87 | ||
BC006730.1 | - | 9 - 10 | 1431 | 3962441 | 87 | |
BC007322.1 | - | 10 - 11 | 1793 | 87 | ||
BC056456.1 | - | 10 - 11 | 1929 | 87 | ||
U48830.1 | - | 10 - 11 | 1502 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |