Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcsk7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001281934.1 | NP_001268863.1 | 13 - 14 | 2557 | 90 | |
NM_008794.2 | NP_032820.2 | 13 - 14 | 2293 | 90 | ||
NR_104055.1 | 13 - 14 | 2286 | 90 | |||
GenBank mRNA | AK148416.1 | - | 13 - 14 | 1878 | 90 | |
AK151980.1 | - | 13 - 14 | 1880 | 90 | ||
AK165389.1 | - | 13 - 14 | 1899 | 90 | ||
BC006730.1 | - | 12 - 13 | 1794 | 3962441 | 90 | |
BC007322.1 | - | 13 - 14 | 2156 | 90 | ||
BC056456.1 | - | 13 - 14 | 2285 | 90 | ||
U48830.1 | - | 13 - 14 | 1865 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Mus musculus Probe spans exons |