Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Etv4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | DQ832277.1 | - | 5 - 6 | 362 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Mouse Tumor Metastasis Panel Inventoried Probe spans exons |