Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pecam1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK037551.1 | - | 6 - 7 | 1551 | 71 | |
AK084527.1 | - | 5 - 6 | 1141 | 71 | ||
AK169431.1 | - | 4 - 5 | 840 | 71 | ||
BC008519.1 | - | 5 - 6 | 1145 | 3592726 | 71 | |
BC085502.1 | - | 5 - 6 | 947 | 5720681 | 71 | |
L06039.1 | - | 5 - 6 | 1030 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |