Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037722.2 | NP_001032811.2 | 15 - 16 | 2046 | 98 | |
NM_009614.2 | NP_033744.1 | 15 - 16 | 2046 | 98 | ||
GenBank mRNA | AF006196.1 | - | 15 - 16 | 2007 | 98 | |
AK039384.1 | - | 15 - 16 | 2026 | 98 | ||
AK048901.1 | - | 15 - 16 | 2033 | 98 | ||
AK149684.1 | - | 15 - 16 | 2028 | 98 | ||
AK149796.1 | - | 2019 | 98 | |||
AK151804.1 | - | 15 - 16 | 1982 | 98 | ||
AK152725.1 | - | 15 - 16 | 1982 | 98 | ||
AK152973.1 | - | 15 - 16 | 1984 | 98 | ||
AK170101.1 | - | 15 - 16 | 2012 | 98 | ||
AK170365.1 | - | 15 - 16 | 2026 | 98 | ||
AK206889.1 | - | 2 - 3 | 310 | 98 | ||
AK218324.1 | - | 1 - 2 | 162 | 98 | ||
BC057909.1 | - | 11 - 12 | 2597 | 98 | ||
EF506571.1 | - | 9 - 10 | 1307 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |