Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adamts1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009621.4 | NP_033751.3 | 1 - 2 | 1139 | 98 | |
GenBank mRNA | AK156830.1 | - | 1 - 2 | 1138 | 98 | |
AK163622.1 | - | 1 - 2 | 1138 | 98 | ||
BC003269.1 | - | 1 - 2 | 1124 | 98 | ||
BC040382.1 | - | 1 - 2 | 1123 | 3156736 | 98 | |
BC050834.1 | - | 1 - 2 | 1123 | 6505494 | 98 | |
D67076.1 | - | 1 - 2 | 1123 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |