Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankfy1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009671.5 | NP_033801.4 | 12 - 13 | 1726 | 109 | |
GenBank mRNA | AB011370.2 | - | 12 - 13 | 1773 | 109 | |
AB098329.1 | - | 12 - 13 | 1724 | 109 | ||
BC139231.1 | - | 12 - 13 | 1737 | 8862253 | 109 | |
BC145583.1 | - | 12 - 13 | 1737 | 9054149 | 109 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |