Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfatc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037177.2 | NP_001032254.1 | 1 - 2 | 303 | 94 | |
NM_001136073.2 | NP_001129545.1 | 1 - 2 | 303 | 94 | ||
NM_010899.3 | NP_035029.2 | 1 - 2 | 303 | 94 | ||
GenBank mRNA | AF289078.1 | - | 1 - 2 | 129 | 94 | |
AK081853.1 | - | 1 - 2 | 270 | 94 | ||
AK135414.1 | - | 1 - 2 | 304 | 94 | ||
BC064006.1 | - | 1 - 2 | 222 | 30290121 | 94 | |
CB590507.1 | - | 246 | 94 | |||
EU887582.1 | - | 1 - 2 | 129 | 94 | ||
EU887588.1 | - | 1 - 2 | 129 | 94 | ||
EU887594.1 | - | 1 - 2 | 129 | 94 | ||
EU887600.1 | - | 1 - 2 | 129 | 94 | ||
U36575.1 | - | 1 - 2 | 199 | 94 | ||
U36576.1 | - | 1 - 2 | 199 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |