Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ntn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010947.3 | NP_035077.1 | 1 - 2 | 1330 | 56 | |
GenBank mRNA | AF128866.1 | - | 1 - 2 | 1170 | 56 | |
AF149094.1 | - | 1 - 2 | 966 | 56 | ||
AF152418.1 | - | 1 - 2 | 1085 | 56 | ||
AK031199.1 | - | 1 - 2 | 1475 | 56 | ||
AK141856.1 | - | 1 - 2 | 1241 | 56 | ||
BC094362.1 | - | 1 - 2 | 1330 | 6830466 | 56 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |