Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdcd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168491.1 | NP_001161963.1 | 5 - 6 | 760 | 107 | |
NM_001168492.1 | NP_001161964.1 | 5 - 6 | 760 | 107 | ||
NM_011050.4 | NP_035180.2 | 5 - 6 | 848 | 107 | ||
GenBank mRNA | AK134366.1 | - | 5 - 6 | 849 | 107 | |
AK172654.1 | - | 5 - 6 | 716 | 107 | ||
BC055739.1 | - | 5 - 6 | 695 | 6414089 | 107 | |
CT010188.1 | - | 4 - 5 | 558 | 107 | ||
D50465.1 | - | 4 - 5 | 628 | 107 | ||
D86344.1 | - | 5 - 6 | 644 | 107 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |