Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Padi4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011061.2 | NP_035191.2 | 13 - 14 | 1575 | 148 | |
GenBank mRNA | AB013850.1 | - | 13 - 14 | 1568 | 148 | |
AK137625.1 | - | 13 - 14 | 1578 | 148 | ||
AK169844.1 | - | 1 - 2 | 200 | 147 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |