Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pygm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011224.1 | NP_035354.1 | 1 - 2 | 296 | 66 | |
GenBank mRNA | AF124787.1 | - | 1 - 2 | 296 | 66 | |
BC012961.1 | - | 1 - 2 | 302 | 3989941 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |