Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1961913 |
BC012961.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1961913 |
NM_011224.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1961913 |
AF124787.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1961913 |
AK019077.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
386088 |
BC145759.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
386088 |
NM_001085351.1 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
386088 |
XM_001471898.1 |
refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
829556 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
438640 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
569423 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
506644 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
30028 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
137517 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
37022 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
105600 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P135468 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1961913 |
BC012961.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1961913 |
NM_011224.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1961913 |
AF124787.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1961913 |
AK019077.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P458973 |
BC012961.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P458973 |
NM_011224.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P458973 |
AF124787.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P458973 |
AK019077.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31934371 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31934454 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31934384 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31934383 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31934447 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31934377 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31934345 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31934364 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31934366 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31934347 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31934373 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31934368 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31934387 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31934374 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31934356 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31934354 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31934349 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31934452 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31934355 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31934448 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31934446 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31934362 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31934359 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31934441 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31934444 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31934445 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31934353 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31934351 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31934457 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31934450 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31934381 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P09096290 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P135468 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P112828 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P458973 |
BC012961.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P458973 |
NM_011224.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P458973 |
AF124787.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P458973 |
AK019077.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P134664 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P135468 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P112828 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P14510334 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P14510350 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P14510327 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P14510354 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934519 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934487 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934376 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934520 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934394 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934401 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934359 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934474 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934452 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934350 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934337 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934479 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934421 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934353 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934498 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934406 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934434 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934508 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934387 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934466 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934493 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934502 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934349 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934492 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934514 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934392 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934336 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934458 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934419 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934480 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934407 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934396 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934488 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934525 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934422 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934454 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934513 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934383 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934384 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934342 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934475 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934429 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934459 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934378 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934484 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934404 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934417 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934390 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934464 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934494 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934471 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934482 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934499 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934413 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934388 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934354 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934381 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934420 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934483 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934399 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934450 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934371 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934522 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934409 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934504 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934366 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934470 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934446 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934501 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934400 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934362 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934515 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934486 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934389 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934472 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934490 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934351 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934510 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934402 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934364 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934408 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934410 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934461 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934517 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934340 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934431 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934478 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934524 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934374 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934368 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31934355 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31934342 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14214777 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31934387 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31934450 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31934422 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31934378 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14472130 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31934347 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31934394 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31934349 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14216910 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31934354 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31934362 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14210554 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14259878 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14510332 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14510343 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31934408 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14510335 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31934413 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31934337 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31934353 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31934367 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14510330 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14460831 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31934388 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14510334 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31934421 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31934442 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31934419 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14452213 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31934351 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31934417 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14463623 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31934345 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31934355 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14510340 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31934396 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31934402 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31934344 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31934405 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31934420 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31934384 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31934389 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31934368 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31934340 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31934404 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14302922 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31934374 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31934409 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14273990 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31934454 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31934452 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31934446 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31934407 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31934401 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31934348 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31934410 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31934359 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31934392 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31934399 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14389878 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31934381 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31934439 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31934436 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14214772 |
- |
- |