Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plod3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011962.3 | NP_036092.1 | 10 - 11 | 1805 | 132 | |
GenBank mRNA | AF046783.1 | - | 10 - 11 | 1213 | 132 | |
AK013195.1 | - | 10 - 11 | 1351 | 132 | ||
AK033360.1 | - | 10 - 11 | 1350 | 132 | ||
AK053320.1 | - | 10 - 11 | 1447 | 132 | ||
AK088948.1 | - | 10 - 11 | 1222 | 132 | ||
AK157741.1 | - | 9 - 10 | 1455 | 132 | ||
AK183180.1 | - | 2 - 3 | 157 | 133 | ||
BC043047.1 | - | 10 - 11 | 1804 | 5703967 | 132 | |
BC054734.1 | - | 10 - 11 | 1805 | 5720686 | 132 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |