Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anxa11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013469.2 | NP_038497.2 | 7 - 8 | 1003 | 69 | |
GenBank mRNA | AK044203.1 | - | 7 - 8 | 976 | 69 | |
AK137158.1 | - | 6 - 7 | 1313 | 69 | ||
AK140375.1 | - | 7 - 8 | 976 | 69 | ||
AK170850.1 | - | 5 - 6 | 2387 | 69 | ||
BC012875.1 | - | 7 - 8 | 923 | 3500318 | 69 | |
CT010331.1 | - | 6 - 7 | 854 | 69 | ||
U65986.1 | - | 7 - 8 | 988 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |