Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bop1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013481.1 | NP_038509.1 | 10 - 11 | 1295 | 79 | |
GenBank mRNA | AK079627.1 | - | 10 - 11 | 1275 | 79 | |
AK129058.1 | - | 10 - 11 | 1268 | 79 | ||
AK144095.1 | - | 10 - 11 | 1306 | 79 | ||
AK167110.1 | - | 10 - 11 | 1278 | 79 | ||
AK167225.1 | - | 10 - 11 | 1279 | 79 | ||
AK167829.1 | - | 10 - 11 | 1273 | 79 | ||
AK167878.1 | - | 10 - 11 | 1283 | 79 | ||
AK168127.1 | - | 10 - 11 | 1276 | 79 | ||
BC012693.1 | - | 10 - 11 | 1269 | 4193123 | 79 | |
U77415.1 | - | 10 - 11 | 1295 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |