Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcsk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013628.2 | NP_038656.1 | 4 - 5 | 622 | 137 | |
GenBank mRNA | AK043742.1 | - | 4 - 5 | 694 | 137 | |
BC108982.1 | - | 4 - 5 | 600 | 40047652 | 137 | |
BC108983.1 | - | 4 - 5 | 600 | 40047655 | 137 | |
M58589.1 | - | 4 - 5 | 622 | 137 | ||
M69196.1 | - | 4 - 5 | 728 | 137 | ||
X57088.1 | - | 4 - 5 | 734 | 137 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Mus musculus Probe spans exons |