Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700027938.mm.1 |
NM_013643.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700027938.mm.1 |
NM_013643.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700027938.mm.1 |
TC0700003050.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700027940.mm.1 |
NM_013643.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700027940.mm.1 |
NM_013643.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700027940.mm.1 |
TC0700003050.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700027925.mm.1 |
NM_001163565.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700027925.mm.1 |
NM_001163565.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700027925.mm.1 |
NM_013643.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700027925.mm.1 |
NM_013643.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700027925.mm.1 |
TC0700003050.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700027936.mm.1 |
NM_013643.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700027936.mm.1 |
NM_013643.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700027936.mm.1 |
TC0700003050.mm.1 |
MTA TC |