Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptdss2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013782.4 | NP_038810.2 | 2 - 3 | 329 | 100 | |
GenBank mRNA | AF099053.1 | - | 2 - 3 | 213 | 100 | |
AK021249.1 | - | 2 - 3 | 294 | 100 | ||
AK031906.1 | - | 2 - 3 | 315 | 100 | ||
AK032686.1 | - | 2 - 3 | 294 | 100 | ||
AK036368.1 | - | 2 - 3 | 330 | 100 | ||
AK048026.1 | - | 2 - 3 | 312 | 100 | ||
AK081275.1 | - | 2 - 3 | 296 | 100 | ||
BC053517.1 | - | 2 - 3 | 266 | 6315175 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |