Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdcd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016688.2 | NP_057897.2 | 2 - 3 | 1074 | 71 | |
GenBank mRNA | AF083929.1 | - | 2 - 3 | 1068 | 71 | |
AK155050.1 | - | 2 - 3 | 1039 | 71 | ||
BC022772.1 | - | 1 - 2 | 111 | 71 | ||
BU518553.1 | - | 2 - 3 | 488 | 71 | ||
DV656988.1 | - | 1 - 2 | 87 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |