Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plrg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016784.3 | NP_058064.2 | 2 - 3 | 226 | 57 | |
GenBank mRNA | AF044334.1 | - | 1 - 2 | 118 | 57 | |
AK075993.1 | - | 2 - 3 | 214 | 57 | ||
AK076007.1 | - | 2 - 3 | 204 | 57 | ||
AK077197.1 | - | 2 - 3 | 227 | 57 | ||
AK080068.1 | - | 2 - 3 | 233 | 57 | ||
BC006750.1 | - | 2 - 3 | 196 | 3709134 | 57 | |
BC052895.1 | - | 2 - 3 | 185 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |