Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Banp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285981.1 | NP_001272910.1 | 8 - 9 | 1306 | 78 | |
NM_016812.4 | NP_058092.2 | 9 - 10 | 1423 | 78 | ||
GenBank mRNA | AF091234.1 | - | 8 - 9 | 1217 | 78 | |
AF235503.1 | - | 9 - 10 | 1334 | 78 | ||
AK179949.1 | - | 2 - 3 | 188 | 78 | ||
AK209622.1 | - | 2 - 3 | 188 | 78 | ||
BC013339.1 | - | 2 - 3 | 279 | 78 | ||
BC021650.1 | - | 9 - 10 | 1341 | 3660187 | 78 | |
BC022168.1 | - | 9 - 10 | 1341 | 4189797 | 78 | |
BC062641.1 | - | 9 - 10 | 1368 | 5718330 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |