Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pla2g6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199023.1 | NP_001185952.1 | 3 - 4 | 1205 | 67 | |
NM_001199024.1 | NP_001185953.1 | 3 - 4 | 1205 | 67 | ||
NM_001199025.1 | NP_001185954.1 | 4 - 5 | 710 | 67 | ||
NM_016915.4 | NP_058611.1 | 3 - 4 | 751 | 67 | ||
GenBank mRNA | AF259401.1 | - | 2 - 3 | 485 | 67 | |
AK144495.1 | - | 4 - 5 | 584 | 67 | ||
AK153333.1 | - | 3 - 4 | 1011 | 67 | ||
BC003487.1 | - | 3 - 4 | 544 | 2811357 | 67 | |
BC049778.1 | - | 3 - 4 | 931 | 67 | ||
BC052845.1 | - | 3 - 4 | 576 | 30054573 | 67 | |
BC057209.1 | - | 3 - 4 | 798 | 5329147 | 67 | |
CF531790.1 | - | 3 - 4 | 634 | 67 | ||
U88624.2 | - | 2 - 3 | 427 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |