Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B4galt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253381.1 | NP_001240310.1 | 6 - 7 | 1362 | 59 | |
NM_017377.5 | NP_059073.1 | 6 - 7 | 1327 | 59 | ||
NR_045570.1 | 7 - 8 | 1382 | 59 | |||
GenBank mRNA | AB019541.2 | - | 4 - 5 | 866 | 59 | |
AF142670.1 | - | 5 - 6 | 971 | 59 | ||
AK145250.1 | - | 7 - 8 | 1384 | 59 | ||
AK165829.1 | - | 6 - 7 | 1321 | 59 | ||
BC138567.1 | - | 5 - 6 | 963 | 8861587 | 59 | |
BC138569.1 | - | 5 - 6 | 963 | 8861589 | 59 | |
BC145335.1 | - | 5 - 6 | 998 | 9053891 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |