Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asah2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018830.1 | NP_061300.1 | 9 - 10 | 1181 | 69 | |
GenBank mRNA | AB037111.1 | - | 10 - 11 | 1466 | 69 | |
AB037181.1 | - | 9 - 10 | 1181 | 69 | ||
AK034493.1 | - | 9 - 10 | 1412 | 69 | ||
AK041937.1 | - | 9 - 10 | 1370 | 69 | ||
AK046540.1 | - | 9 - 10 | 1375 | 69 | ||
AK047692.1 | - | 9 - 10 | 1369 | 69 | ||
AK080951.1 | - | 9 - 10 | 1358 | 69 | ||
AK166100.1 | - | 9 - 10 | 1429 | 69 | ||
BC137898.1 | - | 9 - 10 | 1164 | 8860917 | 69 | |
BC145016.1 | - | 9 - 10 | 1167 | 9053564 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |